missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride, 95%, Maybridge

Brand:  Maybridge CC68574DA

Code : 31

Additional Details : CAS Number : 944450-92-4 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10507674

  • £97.00 / 1g


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride
94.5
95%
C11H16ClNO·ClH
MFCD10700059
944450-92-4
100.0
95%
250.17

Specifications

4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride
94.5
Amber Glass Bottle
95%
C11H16ClNO·ClH
MFCD10700059
944450-92-4
100.0
95%
1g
250.17
Videos
SDS