missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromandelic acid, 95%, Thermo Scientific™

Catalog Number p-3212047
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
492-86-4
MFCD00042724
BWSFWXSSALIZAU-UHFFFAOYSA-N
4-chloromandelic acid, 2-4-chlorophenyl-2-hydroxyacetic acid, p-chloromandelic acid, p-chloro mandelic acid, 4-chloro-dl-mandelic acid, 4-chloromandelic acid, pract., benzeneacetic acid, 4-chloro-.alpha.-hydroxy, dl-4-chloromandelic acid, mandelic acid, p-chloro, 4-chlorophenyl hydroxy acetic acid
10299
2-(4-chlorophenyl)-2-hydroxyacetic acid
C1=CC(=CC=C1C(C(=O)O)O)Cl
This item is not returnable. View return policy
492-86-4
MFCD00042724
BWSFWXSSALIZAU-UHFFFAOYSA-N
4-chloromandelic acid, 2-4-chlorophenyl-2-hydroxyacetic acid, p-chloromandelic acid, p-chloro mandelic acid, 4-chloro-dl-mandelic acid, 4-chloromandelic acid, pract., benzeneacetic acid, 4-chloro-.alpha.-hydroxy, dl-4-chloromandelic acid, mandelic acid, p-chloro, 4-chlorophenyl hydroxy acetic acid
10299
2-(4-chlorophenyl)-2-hydroxyacetic acid
C1=CC(=CC=C1C(C(=O)O)O)Cl
Melting Point 115.0°C to 120.0°C
CAS Min % 94.0
Color White to Yellow
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 94% min. (HPLC)
Molecular Formula C8H7ClO3
Linear Formula ClC6H4CH(OH)COOH
Molecular Weight (g/mol) 186.59
Formula Weight 186.59
Percent Purity 95%
Chemical Name or Material 4-Chloromandelic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloromandelic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'