missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorofuro[3,2-c]pyridine, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 153.57 |
Percent Purity | 95% |
Chemical Name or Material | 4-Chlorofuro[3, 2-c]pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'