missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals

Catalog Number p-86849
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
1615-02-7
C9H7ClO2
182.60
MFCD00004396
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
637797
CHEBI:61116
(E)-3-(4-chlorophenyl)prop-2-enoic acid
OC(=O)\C=C\C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
1615-02-7
C9H7ClO2
182.60
MFCD00004396
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
637797
CHEBI:61116
(E)-3-(4-chlorophenyl)prop-2-enoic acid
OC(=O)\C=C\C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 246°C to 250°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClC6H4CH=CHCO2H
Quantity 25 g
Beilstein 09, 594
Formula Weight 182.61
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Chlorocinnamic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorocinnamic acid, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'