missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chlorobutyryl chloride, 98%

Brand:  Alfa Aesar™ A12249.09

Code : NEW

Additional Details : CAS Number : 4635-59-0 Weight : 0.01000kg Transport : UN number : 2927 Chem class : 6.1 Pack group : II

Quantity 10g

 View more versions of this product

Product Code. 11324749

  • £9.70 / 10g


EU Stock 23
Add to basket

Description

Description

Applications
4-Chlorobutyryl chloride (4-CBCl) is mainly used in the production of pharmaceuticals and agrochemicals. It is used as pyrazinamide intermediate.

Solubility
soluble in diethyl ether, ethanol, slightly soluble in water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agent, bases,
Specifications

Chemical Identifiers

4-Chlorobutyryl chloride
C4H6Cl2O
141
UN2927
4635-59-0
ClCH2CH2CH2COCl
MFCD00000754
773860

Specifications

4-Chlorobutyryl chloride
173° to 174°C
10g
Acrid
C4H6Cl2O
141
UN2927
85°C (185°F)
−49°C
4635-59-0
98%
ClCH2CH2CH2COCl
MFCD00000754
773860
Videos
SDS