missing translation for 'onlineSavingsMsg'
Learn More

4-chlorobutyl chloroformate, 97%, ACROS Organics™

Brand:  Acros Organics 314490250

Code : 29

Additional Details : CAS Number : 37693-18-8 Weight : 0.08100kg Transport : UN number : 3277 Chem class : 6.1 Pack group : II

Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10762243

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Coupling reagent
Specifications

Chemical Identifiers

4-Chlorobutyl chloroformate
37693-18-8
100.0
97%
ClCO2(CH2)4Cl
MFCD00134472
97%
96.0
≥96.0%
C5H8Cl2O2
171.02

Specifications

4-Chlorobutyl chloroformate
Clear colorless
97%
96.0
Authentic
1.4520 to 1.4540 (20°C, 589nm)
≥96.0%
C5H8Cl2O2
171.02
1.2500g/mL
25mL
37693-18-8
100.0
Glass bottle
Liquid
97%
ClCO2(CH2)4Cl
MFCD00134472
Videos
SDS