missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile, 99%, ACROS Organics™

Brand:  Acros Organics 151871000

Code : 29

Additional Details : CAS Number : 623-03-0 Weight : 0.15000kg

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10705991

  • £46.70 / 100g


Estimated Shipment
14-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chlorobenzonitrile
98.5
≥98.5%
C7H4ClN
137.57
PCBN
623-03-0
100.0
99%
ClC6H4CN
MFCD00001813

Specifications

4-Chlorobenzonitrile
91° to 94°C
623-03-0
100.0
Plastic bottle
Flakes or Chunks
99%
ClC6H4CN
MFCD00001813
PCBN
White to yellow
100g
98.5
Authentic
0.1% max.
≥98.5%
C7H4ClN
137.57
09, 341
Videos
SDS