missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-85587
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
74-11-3
C7H5ClO2
156.57
MFCD00002531
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
6318
CHEBI:30747
4-chlorobenzoic acid
C1=CC(=CC=C1C(=O)O)Cl
This item is not returnable. View return policy
74-11-3
C7H5ClO2
156.57
MFCD00002531
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
6318
CHEBI:30747
4-chlorobenzoic acid
C1=CC(=CC=C1C(=O)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 240.0°C to 243.0°C
Color White to Yellow
Boiling Point 274.0°C to 276.0°C
Flash Point 238°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H4CO2H
Quantity 25 g
Beilstein 09, 340
Merck Index 15, 2126
Solubility Information Solubility in water: 0,08g/L (20°C). Other solubilities: soluble in hot water, freely soluble in alcohol and ether
Formula Weight 156.57
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'