missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzene-1-carboximidamide hydrochloride, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Linear Formula | ClC6H4C(=NH)NH2.HCl |
Formula Weight | 191.06 |
Percent Purity | 95% |
Quantity | 10 g |
Packaging | Amber Glass Bottle |
Chemical Name or Material | 4-Chlorobenzene-1-carboximidamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'