missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamidine hydriodide, 96%, Thermo Scientific Chemicals

Catalog Number p-7081554
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
115297-57-9
C7H8ClIN2
282.509
MFCD00051987
YMEVGWRZEYGNJO-UHFFFAOYSA-N
4-chlorobenzamidine hydroiodide, 4-chlorobenzimidamide hydroiodide, 4-chlorobenzamidine hydriodide, chlorophenamidine hydroiodide, 4-chlorobenzenecarboximidamide hydroiodide, acmc-1bu33, benzenecarboximidamide,4-chloro-,hydriodide 1:1, benzenecarboximidamide,4-chloro-, hydriodide 1:1
2734221
4-chlorobenzenecarboximidamide;hydroiodide
C1=CC(=CC=C1C(=N)N)Cl.I
This item is not returnable. View return policy
115297-57-9
C7H8ClIN2
282.509
MFCD00051987
YMEVGWRZEYGNJO-UHFFFAOYSA-N
4-chlorobenzamidine hydroiodide, 4-chlorobenzimidamide hydroiodide, 4-chlorobenzamidine hydriodide, chlorophenamidine hydroiodide, 4-chlorobenzenecarboximidamide hydroiodide, acmc-1bu33, benzenecarboximidamide,4-chloro-,hydriodide 1:1, benzenecarboximidamide,4-chloro-, hydriodide 1:1
2734221
4-chlorobenzenecarboximidamide;hydroiodide
C1=CC(=CC=C1C(=N)N)Cl.I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼235°C (decomposition)
Quantity 1 g
Beilstein 8280383
Formula Weight 282.51
Percent Purity 96%
Chemical Name or Material 4-Chlorobenzamidine hydriodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'