missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamide, 98+%, Thermo Scientific™

Catalog Number p-85482
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
619-56-7
C7H6ClNO
155.58
MFCD00007993
BLNVISNJTIRAHF-UHFFFAOYSA-N
p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
12084
4-chlorobenzamide
NC(=O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
619-56-7
C7H6ClNO
155.58
MFCD00007993
BLNVISNJTIRAHF-UHFFFAOYSA-N
p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
12084
4-chlorobenzamide
NC(=O)C1=CC=C(Cl)C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 177.0°C to 181.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula ClC6H4CONH2
Quantity 25 g
Beilstein 09, 341
Formula Weight 155.58
Percent Purity ≥98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Chlorobenzamide, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'