missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline, 98%, Thermo Scientific Chemicals

Catalog Number 10355930
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
106-47-8
C6H6ClN
127.57
MFCD00007835
QSNSCYSYFYORTR-UHFFFAOYSA-N
p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
7812
CHEBI:20331
4-chloroaniline
C1=CC(=CC=C1N)Cl
This item is not returnable. View return policy
106-47-8
C6H6ClN
127.57
MFCD00007835
QSNSCYSYFYORTR-UHFFFAOYSA-N
p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
7812
CHEBI:20331
4-chloroaniline
C1=CC(=CC=C1N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 68.0°C to 72.0°C
CAS Min % 97.5
Color Beige to Brown-Purple
CAS Max % 100.0
Boiling Point 232.0°C
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4NH2
Viscosity 1.2 mPa.s (90°C)
Beilstein 12,607
Merck Index 15,2118
Solubility Information Solubility in water: 3g/L in water (20°C). Other solubilities: freely soluble in alcohol, ether, acetone and, carbon disulfide
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 4-Chloroaniline, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'