4'-Chloroacetoacetanilide 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 158550050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 101-92-8 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 10333382

Quantity Price
1 £ 9.7 / 5g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4'-Chloroacetoacetanilide
Name Note 98%
CAS 101-92-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10ClNO2
Linear Formula CH3COCH2CONHC6H4Cl
Formula Weight 211.65
MDL Number MFCD00000613

Specification

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 211.65
Quantity 5g
Color Off-white to beige
CAS Min % 97.5
EINECS Number 202-989-6
Linear Formula CH3COCH2CONHC6H4Cl
Infrared Spectrum Authentic
CAS 101-92-8
MDL Number MFCD00000613
RTECS Number AK4375000
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H10ClNO2
Name Note 98%
CAS Max % 100.0
TSCA TSCA
Chemical Name or Material 4'-Chloroacetoacetanilide
Melting Point 129° to 133°C
Documentation

Technical documents