missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N,N-diethylbenzamide, 97%, Thermo Scientific™

Catalog Number 15415558
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
7461-38-3
C11H14ClNO
211.689
MFCD00584080
VQIFBGNOSBKNJI-UHFFFAOYSA-N
benzamide, 4-chloro-n,n-diethyl, benzamide nsc 404988
346696
4-chloro-N,N-diethylbenzamide
CCN(CC)C(=O)C1=CC=C(C=C1)Cl
This item is not returnable. View return policy
7461-38-3
C11H14ClNO
211.689
MFCD00584080
VQIFBGNOSBKNJI-UHFFFAOYSA-N
benzamide, 4-chloro-n,n-diethyl, benzamide nsc 404988
346696
4-chloro-N,N-diethylbenzamide
CCN(CC)C(=O)C1=CC=C(C=C1)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Formula Weight 211.69
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Chloro-N,N-diethylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'