missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N'-hydroxybenzenecarboximidamide, 95%, Thermo Scientific™

Catalog Number p-2706239
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
5033-28-3
C7H7ClN2O
170.60
MFCD00029674
QBGONPQFBDUVPG-UHFFFAOYSA-N
z-4-chloro-n'-hydroxybenzamidine, 4-chloro-n-hydroxy-benzamidine, 4-chloro-n-hydroxybenzenecarboximidamide, 4-chlorobenzamide oxime, z-4-chloro-n'-hydroxybenzimidamide, 4-chloro-n'-hydroxybenzene-1-carboximidamide, z-4-chloro-n'-hydroxybenzene-1-carboximidamide, 4-chloro-n'-hydroxybenzimidamide, 4-chloro-benzamidoxime, 4-chlorophenyl hydroxyimino methylamine
9561067
(Z)-4-chloro-N'-hydroxybenzene-1-carboximidamide
N\C(=N/O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
5033-28-3
C7H7ClN2O
170.60
MFCD00029674
QBGONPQFBDUVPG-UHFFFAOYSA-N
z-4-chloro-n'-hydroxybenzamidine, 4-chloro-n-hydroxy-benzamidine, 4-chloro-n-hydroxybenzenecarboximidamide, 4-chlorobenzamide oxime, z-4-chloro-n'-hydroxybenzimidamide, 4-chloro-n'-hydroxybenzene-1-carboximidamide, z-4-chloro-n'-hydroxybenzene-1-carboximidamide, 4-chloro-n'-hydroxybenzimidamide, 4-chloro-benzamidoxime, 4-chlorophenyl hydroxyimino methylamine
9561067
(Z)-4-chloro-N'-hydroxybenzene-1-carboximidamide
N\C(=N/O)C1=CC=C(Cl)C=C1
CAS Min % 95.0
Assay Percent Range 95%
Linear Formula ClC6H4C(=NOH)NH2
Formula Weight 170.6
Percent Purity 95%
Quantity 250 mg
Chemical Name or Material 4-Chloro-N'-hydroxybenzenecarboximidamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'