missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-cyclopentyl-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65458.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472559

  • £74.73 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-cyclopentyl-3-sulfamoylbenzamide
302.78
MFCD10931676

Specifications

4-Chloro-N-cyclopentyl-3-sulfamoylbenzamide
Solid
302.78
1g
95%
MFCD10931676
Videos
SDS