missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(5-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59950.03

Code : NEW

Additional Details : CAS Number : 528835-13-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466408

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(5-fluoro-2-methylphenyl)benzamide
528835-13-4
263.7
MFCD03558380

Specifications

4-Chloro-N-(5-fluoro-2-methylphenyl)benzamide
1g
97%
MFCD03558380
528835-13-4
Solid
263.7
Videos
SDS