missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(5-fluoro-2-methylphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65806.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429916

  • £134.62 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(5-fluoro-2-methylphenyl)-3-sulfamoylbenzamide
342.77
MFCD06675367

Specifications

4-Chloro-N-(5-fluoro-2-methylphenyl)-3-sulfamoylbenzamide
Solid
342.77
1g
95%
MFCD06675367
Videos
SDS