Alfa Aesar™ 4-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65517.MD

250MG 4-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide, 95% 250mg

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15425755

Quantity Price
1 £ 23.0 / 250mg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide
Formula Weight 353.77
MDL Number MFCD27975775

Specification

Formula Weight 353.77
Chemical Name or Material 4-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide
Physical Form Solid
Assay 95%
MDL Number MFCD27975775
Quantity 250mg