missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-iodophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59989.03

Code : NEW

Additional Details : CAS Number : 299954-64-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416458

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(4-iodophenyl)benzamide
299954-64-6
357.58
MFCD00554075

Specifications

4-Chloro-N-(4-iodophenyl)benzamide
1g
Light Sensitive
97%
MFCD00554075
299954-64-6
White
Solid
357.58
Videos
SDS