missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Chloro-N-(4-fluorophenyl)-3-nitrobenzamide, 97%
Brand: Alfa Aesar™ H59934.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Chloro-N-(4-fluorophenyl)-3-nitrobenzamide | |
294.67 | |
MFCD00594545 |
Specifications
4-Chloro-N-(4-fluorophenyl)-3-nitrobenzamide | |
Solid | |
294.67 |
1g | |
97% | |
MFCD00594545 |