missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-chlorobenzyl)benzamide, 97%

Brand:  Alfa Aesar™ H59336.03

Code : NEW

Additional Details : CAS Number : 130780-10-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475608

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(4-chlorobenzyl)benzamide
130780-10-8
280.15
MFCD01011332

Specifications

4-Chloro-N-(4-chlorobenzyl)benzamide
1g
97%
MFCD01011332
130780-10-8
Solid
280.15
Videos
SDS