missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-[4-chloro-3-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65474.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409676

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

4-Chloro-N-[4-chloro-3-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide
413.2
MFCD07338243

Specifications

4-Chloro-N-[4-chloro-3-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide
Solid
413.2
1g
95%
MFCD07338243
Videos
SDS