missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-chloro-2-fluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59154.03

Code : NEW

Additional Details : CAS Number : 545362-76-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465378

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(4-chloro-2-fluorophenyl)benzamide
545362-76-3
284.12
MFCD03389479

Specifications

4-Chloro-N-(4-chloro-2-fluorophenyl)benzamide
1g
Solid
284.12
545362-76-3
Off-white
97%
MFCD03389479
Videos
SDS