missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-fluorophenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65649.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409806

  • £99.64 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3-fluorophenyl)-3-sulfamoylbenzamide
328.75
MFCD27975797

Specifications

4-Chloro-N-(3-fluorophenyl)-3-sulfamoylbenzamide
Solid
328.75
1g
95%
MFCD27975797
Videos
SDS