missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-fluoro-4-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59344.03

Code : NEW

Additional Details : CAS Number : 328026-95-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495618

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3-fluoro-4-methylphenyl)benzamide
328026-95-5
263.7
MFCD02603478

Specifications

4-Chloro-N-(3-fluoro-4-methylphenyl)benzamide
1g
97%
MFCD02603478
328026-95-5
Solid
263.7
Videos
SDS