missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59912.03

Code : NEW

Additional Details : CAS Number : 528844-84-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476358

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3-fluoro-2-methylphenyl)benzamide
528844-84-0
263.7
MFCD03558817

Specifications

4-Chloro-N-(3-fluoro-2-methylphenyl)benzamide
1g
Solid
263.7
528844-84-0
Off-white
97%
MFCD03558817
Videos
SDS