missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-chloro-4-methoxyphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59645.03

Code : NEW

Additional Details : CAS Number : 449166-29-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465156

  • £358.49 / 1g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3-chloro-4-methoxyphenyl)benzamide
449166-29-4
296.15
MFCD01608945

Specifications

4-Chloro-N-(3-chloro-4-methoxyphenyl)benzamide
1g
97%
MFCD01608945
449166-29-4
Solid
296.15
Videos
SDS