missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-chloro-4-methoxyphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65048.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489336

  • £131.44 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3-chloro-4-methoxyphenyl)-3-sulfamoylbenzamide
375.23
MFCD27975715

Specifications

4-Chloro-N-(3-chloro-4-methoxyphenyl)-3-sulfamoylbenzamide
Solid
375.23
1g
95%
MFCD27975715
Videos
SDS