missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-chloro-4-fluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59715.03

Code : NEW

Additional Details : CAS Number : 256444-32-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486108

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3-chloro-4-fluorophenyl)benzamide
256444-32-3
284.12
MFCD01182212

Specifications

4-Chloro-N-(3-chloro-4-fluorophenyl)benzamide
1g
97%
MFCD01182212
256444-32-3
Solid
284.12
Videos
SDS