missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3-chloro-2-methylphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65930.03

UNSPSC : 12352106

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15490017

  • £99.64 / 1g


Estimated Shipment date
from Supplier 12-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

4-Chloro-N-(3-chloro-2-methylphenyl)-3-sulfamoylbenzamide
359.23
MFCD28098843

Specifications

4-Chloro-N-(3-chloro-2-methylphenyl)-3-sulfamoylbenzamide
Solid
359.23
1g
95%
MFCD28098843
Documentation

Technical Documents