missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,5-dimethylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59423.03

Code : NEW

Additional Details : CAS Number : 127292-04-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485738

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3,5-dimethylphenyl)benzamide
127292-04-0
259.74
MFCD00783850

Specifications

4-Chloro-N-(3,5-dimethylphenyl)benzamide
1g
Solid
259.74
127292-04-0
Orange
97%
MFCD00783850
Videos
SDS