missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,5-dimethoxyphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59060.03

Code : NEW

Additional Details : CAS Number : 312922-57-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415248

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3,5-dimethoxyphenyl)benzamide
312922-57-9
291.73
MFCD00429144

Specifications

4-Chloro-N-(3,5-dimethoxyphenyl)benzamide
1g
97%
MFCD00429144
312922-57-9
Solid
291.73
Videos
SDS