missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,5-difluorophenyl)butyramide, 97%

Brand:  Alfa Aesar™ H59339.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415618

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3,5-difluorophenyl)butyramide
233.64
MFCD09728965

Specifications

4-Chloro-N-(3,5-difluorophenyl)butyramide
Solid
233.64
1g
97%
MFCD09728965
Videos
SDS