missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,5-difluorophenyl)-3-nitrobenzamide, 97%

Brand:  Alfa Aesar™ H59733.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495226

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3,5-difluorophenyl)-3-nitrobenzamide
312.66
MFCD07082022

Specifications

4-Chloro-N-(3,5-difluorophenyl)-3-nitrobenzamide
Solid
312.66
1g
97%
MFCD07082022
Videos
SDS