missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(3,4-difluorophenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65652.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469806

  • £91.90 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(3,4-difluorophenyl)-3-sulfamoylbenzamide
346.74
MFCD27975799

Specifications

4-Chloro-N-(3,4-difluorophenyl)-3-sulfamoylbenzamide
Solid
346.74
1g
95%
MFCD27975799
Videos
SDS