Learn More
Alfa Aesar™ 4-Chloro-N-(3,4-dichlorophenyl)benzamide, 97%
Brand: Alfa Aesar™ H59101.03
Code : NEW
Additional Details : CAS Number : 56661-50-8 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Chloro-N-(3,4-dichlorophenyl)benzamide | |
56661-50-8 | |
300.57 | |
MFCD00018677 |
Specifications
4-Chloro-N-(3,4-dichlorophenyl)benzamide | |
1g | |
97% | |
MFCD00018677 |
56661-50-8 | |
Solid | |
300.57 |