missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-[2-fluoro-5-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65842.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469936

  • £62.96 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-[2-fluoro-5-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide
396.74
MFCD28098831

Specifications

4-Chloro-N-[2-fluoro-5-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide
Solid
396.74
1g
95%
MFCD28098831
Videos
SDS