missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2-fluoro-3-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59242.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475488

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2-fluoro-3-methylphenyl)benzamide
263.7
MFCD22683047

Specifications

4-Chloro-N-(2-fluoro-3-methylphenyl)benzamide
Pale purple
97%
MFCD22683047
1g
Solid
263.7
Videos
SDS