missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2-ethylhexyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65229.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439486

  • £66.46 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2-ethylhexyl)-3-sulfamoylbenzamide
346.87
MFCD27975739

Specifications

4-Chloro-N-(2-ethylhexyl)-3-sulfamoylbenzamide
Solid
346.87
1g
95%
MFCD27975739
Videos
SDS