missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2-ethoxyphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65860.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429956

  • £59.78 / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2-ethoxyphenyl)-3-sulfamoylbenzamide
354.81
MFCD27975832

Specifications

4-Chloro-N-(2-ethoxyphenyl)-3-sulfamoylbenzamide
Solid
354.81
1g
95%
MFCD27975832
Videos
SDS