missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65759.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499876

  • £59.78 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide
413.2
MFCD27975813

Specifications

4-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]-3-sulfamoylbenzamide
Solid
413.2
1g
95%
MFCD27975813
Videos
SDS