missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,5-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59250.03

Code : NEW

Additional Details : CAS Number : 825659-03-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485498

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2,5-difluorophenyl)benzamide
825659-03-8
267.66
MFCD04680216

Specifications

4-Chloro-N-(2,5-difluorophenyl)benzamide
1g
97%
MFCD04680216
825659-03-8
Solid
267.66
Videos
SDS