missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,5-dichlorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59867.03

Code : NEW

Additional Details : CAS Number : 2447-97-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466298

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2,5-dichlorophenyl)benzamide
2447-97-4
300.57
MFCD00018671

Specifications

4-Chloro-N-(2,5-dichlorophenyl)benzamide
1g
97%
MFCD00018671
2447-97-4
Solid
300.57
Videos
SDS