missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,4-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59055.03

Code : NEW

Additional Details : CAS Number : 331435-74-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405238

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2,4-difluorophenyl)benzamide
331435-74-6
267.66
MFCD00751582

Specifications

4-Chloro-N-(2,4-difluorophenyl)benzamide
1g
97%
MFCD00751582
331435-74-6
Solid
267.66
Videos
SDS