Alfa Aesar™ 4-Chloro-N-(2,4-dichlorophenyl)-3-sulfamoylbenzamide, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65208.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15419466

Quantity Price
1 £ 16.64 / 250mg
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Chloro-N-(2,4-dichlorophenyl)-3-sulfamoylbenzamide
Formula Weight 379.65
MDL Number MFCD28098769

Specification

Formula Weight 379.65
Assay 95%
Physical Form Solid
Quantity 250mg
MDL Number MFCD28098769
Chemical Name or Material 4-Chloro-N-(2,4-dichlorophenyl)-3-sulfamoylbenzamide
Documentation

Technical documents