missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,4-dibromophenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65837.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439936

  • £79.71 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2,4-dibromophenyl)-3-sulfamoylbenzamide
468.55
MFCD27975825

Specifications

4-Chloro-N-(2,4-dibromophenyl)-3-sulfamoylbenzamide
Solid
468.55
1g
95%
MFCD27975825
Videos
SDS