missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(2,3-dimethylphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65963.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450047

  • £85.44 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-N-(2,3-dimethylphenyl)-3-sulfamoylbenzamide
338.81
MFCD28098845

Specifications

4-Chloro-N-(2,3-dimethylphenyl)-3-sulfamoylbenzamide
Solid
338.81
1g
95%
MFCD28098845
Videos
SDS