missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-L-phenylalanine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
Solubility
Soluble in acetic acid. Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
Solubility
Soluble in acetic acid. Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 235°C to 238°C |
UN Number | UN2811 |
Beilstein | 2416150 |
Solubility Information | Soluble in acetic acid. Slightly soluble in water. |
IUPAC Name | 2-amino-3-(4-chlorophenyl)propanoic acid |
Formula Weight | 199.63 |
Percent Purity | 97% |
Quantity | 5 g |
Physical Form | Solid |
Chemical Name or Material | 4-Chloro-L-phenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'