missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-DL-phenylglycine, 98%, Thermo Scientific Chemicals

Catalog Number p-7053434
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
6212-33-5
C8H8ClNO2
185.607
MFCD00049328
QGJGBYXRJVIYGA-UHFFFAOYSA-N
2-amino-2-4-chlorophenyl acetic acid, amino 4-chlorophenyl acetic acid, amino-4-chloro-phenyl-acetic acid, dl-2-4-chlorophenyl glycine, 2-4-chlorophenyl glycine, +/--4-chlorophenylglycine, h-dl-phg 4-cl-oh, 4-chloro-dl-phenylglycine, dl-4-chlorophenyl glycine, 4-chlorophenyl glycine
587821
2-amino-2-(4-chlorophenyl)acetic acid
C1=CC(=CC=C1C(C(=O)O)N)Cl
This item is not returnable. View return policy
6212-33-5
C8H8ClNO2
185.607
MFCD00049328
QGJGBYXRJVIYGA-UHFFFAOYSA-N
2-amino-2-4-chlorophenyl acetic acid, amino 4-chlorophenyl acetic acid, amino-4-chloro-phenyl-acetic acid, dl-2-4-chlorophenyl glycine, 2-4-chlorophenyl glycine, +/--4-chlorophenylglycine, h-dl-phg 4-cl-oh, 4-chloro-dl-phenylglycine, dl-4-chlorophenyl glycine, 4-chlorophenyl glycine
587821
2-amino-2-(4-chlorophenyl)acetic acid
C1=CC(=CC=C1C(C(=O)O)N)Cl

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 0.1mol/L NaOH (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Solubility Information Soluble in 0.1mol/L NaOH (almost transparency).
Formula Weight 185.61
Percent Purity 98%
Quantity 25 g
Physical Form Solid
Chemical Name or Material 4-Chloro-DL-phenylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'